This product is now discontinued, View Related Products
Trajan SGE eVol® Digital Analytical Syringe for MEPS (micro SPE)
A faster method for sample separation
  • Use less solvent and reduce waste with microliter needles
  • Smaller volumes process quicker
  • Semiautomated functions minimize operator intervention
This handheld digital syringe features a familiar touch wheel user interface and includes a full-color screen. Use with 5 µL to 1 mL MEPS syringes. Easily and quickly change XCHANGE® enabled syringes. Custom programming function allows process to be validated and semiautomated before transition to a fully automated platform. Extracted samples are compatible with gas and liquid chromatography systems.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Needle Included Syringe Capacity (µL)
Availability Pricing
Trajan SGE EVOL MEPS Electronic Syringe; 500 µL EW-17940-07 Mfr # 2910026
COMPARE
No 500
Call For Price
Trajan SGE EVOL MEPS Electronic Syringe; 50 µL EW-17940-08 Mfr # 2910027
COMPARE
No 50
Call For Price
Trajan SGE EVOL MEPS Electronic Syringe, 100 µL, Removable Needle, 25G; Bevel Tip EW-17940-09 Mfr # 2910028
COMPARE
Yes 100