This product is now discontinued, View Related Products
TriForest Serum Vials
Protect your samples with leakproof vials
  • Ideal for safe storage, transport, and transfer of serums and substrates
  • Constructed of optically clear shatterproof plastic
The polyethylene terephthalate copolyester (PETG) serum vials are presterilized via gamma irradiation, sterile to SAL 10-6. These vials meet current biological and pharmaceutical regulatory standards including USP Class VI, noncytotoxic, nonpyrogenic, USP 661, European Pharmacopeia for Abnormal toxicity, and are Animal Derived Component Free (ADCF). The PETG vials are cleanroom production packaged with a blue tamper-evident polyethylene (PE) cap engaged. The transparent PETG vials feature molded graduations for reference measurements.
The polycarbonate (PC) serum vials and polypropylene (PP) caps come nonsterile with lot number traceability. These can be autoclaved, up to 125°C, and up to 10 autoclave cycles. The temperature range of these vials is -193 to 257°F (-125 to 125°C). The caps are packaged separately.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Cap Type Tube Material Volume (mL)
Availability Pricing
TriForest Serum Vials, Sterile, 5 mL, PETG, 1600/CS EW-06019-33 Mfr # RGC0005S
COMPARE
Tamper Evident Cap PETG 5
Call For Price
TriForest Serum Vials, Sterile, 10 mL, PETG, 1600/CS EW-06019-34 Mfr # RGC0010S
COMPARE
Tamper Evident Cap PETG 10
Call For Price
TriForest Serum Vials, Lot Traceable, 5 mL, PC, 1600/CS EW-06019-35 Mfr # RPC0005
COMPARE
Standard Cap Polycarbonate 5
Call For Price
TriForest Serum Vials, Lot Traceable, 10 mL, PC, 1600/CS EW-06019-36 Mfr # RPC0010
COMPARE
Standard Cap Polycarbonate 10