Troemner Class-F Stainless Steel Test Weight Sets
Stainless Steel Test Weights are primarily used to test commercial weighing devices by state and local weights and measures officials, device installers, and service technicians.
"Small' Weight sets where the total weight is less than 15 lb are supplied in sturdy plastic cases with a carrying handle.
"Large" Weight sets where the total weight is greater than 15 lb are supplied in a case with metal reinforced corners and bottoms with padded handles for easier carrying.
Weights 8 oz and above each have their own compartment within the dense foam interior. Smaller weights are contained within a smaller synthetic leather case that fits into the larger “master” carrying case. This “case-in-a-case” design helps keep smaller weights from getting lost and also allows smaller weights to be taken out of the master case and used closer to the user’s application.
$728.00 - $2,060.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Contents of Set Mass Certification Mass Class
Availability Pricing
Troemner TW-2000-01T Class F Stainless Steel Test Weight Set with NIST Calibration Certificate; 2000 g to 1 mg EW-09108-91 Mfr # TW-2000-01T
COMPARE
2000g, 1000g, 500g, (2) 200g, 100g, 50g, (2) 20g, 10g, 5g, (2) 2g, 1g, 500mg, (2) 200mg, 100mg, 50mg, (2) 20mg, 10mg, 5mg, (2) 2mg, 1mg NIST/NVLAP Class F
$920.00USD / Each
Troemner TW-5 5 lb to 1/32 oz, Class F Stainless Steel Test Weight Set; 14 pieces EW-09108-92 Mfr # TW-5
COMPARE
5lb, (2) 2lb, 1lb, 8oz, 4oz, 2oz, 1oz, 1/2oz, 1/4oz, 1/8oz, 1/16oz, (2) 1/32oz None Class F
$728.00USD / Each
Troemner TW-50 5 lb to 1/32 oz, Class F Stainless Steel Test Weight Set; 20 pieces EW-09108-93 Mfr # TW-50
COMPARE
(5) 5lb, (5) 1lb, 8oz, 4oz, 2oz, 1oz, 1/2oz, 1/4oz, 1/8oz, 1/16oz, (2) 1/32oz None Class F
$1,500.00USD / Each
Troemner TW-5000-01T Class F Stainless Steel Test Weight Set with NIST Calibration Certificate; 5 kg to 1 mg EW-09108-95 Mfr # TW-5000-01T
COMPARE
5000g, (2) 2000g, 1000g, 500g, (2) 200g, 100g, 50g, (2) 20g, 10g, 5g, (2) 2g, 1g, 500mg, (2) 200mg, 100mg, 50mg, (2) 20mg, 10mg, 5mg, (2) 2mg, 1mg NIST/NVLAP Class F
$1,250.00USD / Each
Troemner TW-50T Class F Stainless Steel Grip-Handle Calibration Set with NIST Calibration Certificate; 5 lb (5) to 1/32 oz EW-09108-96 Mfr # TW-50T
COMPARE
(5) 5lb, (5) 1lb, 8oz, 4oz, 2oz, 1oz, 1/2oz, 1/4oz, 1/8oz, 1/16oz, (2) 1/32oz NIST/NVLAP Class F
$1,920.00USD / Each
Troemner TW-55-01T Class F Stainless Steel Grip-Handle Calibration Set NIST Calibration Certificate; 2 kg (5) to 1 mg, 15 kg EW-09108-98 Mfr # TW-55-01T
COMPARE
(5) 2000g, (5) 500g, (2) 200g, 100g, 50g, (2) 20g, 10g, 5g, (2) 2g, 1g, 500mg, (2) 200mg, 100mg, 50mg, (2) 20mg, 10mg, 5mg, (2) 2mg, 1mg NIST/NVLAP Class F
$2,060.00USD / Each
Troemner TW-5T Class F Stainless Steel Test Weight Set with NIST Calibration Certificate; 5 lb to 1/32 oz EW-09108-99 Mfr # TW-5T
COMPARE
5lb, (2) 2lb, 1lb, 8oz, 4oz, 2oz, 1oz, 1/2oz, 1/4oz, 1/8oz, 1/16oz, (2) 1/32oz NIST/NVLAP Class F
$1,230.00USD / Each
Showing 1- 7 of 7
Need Help with this product?
Call Us At1-847-549-7600
Live chat