True Union Three-Way Four-Position Ball Valves
These PVC valves have EPDM O-rings and seats made of PTFE. Connections are true union NPT(F) pipe thread. True union design allows you to separate the valve body without removing pipe connectors—simplifies cleaning and maintenance. Maximum pressure is 150 psi (10.3 bar); maximum temperature is 140°F (60°C). Rotate handle 360° to change flow channel. Valves include both socket and NPT fittings.
$75.60 - $247.70USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cv Port Size (NPT(F))
Availability Pricing
True Union, Three-Way Four-Position Ball Valve, 1/2" NPT(F) EW-98710-00 Mfr # 253850
COMPARE
8 0.5
$85.75USD / Each
053229 True Union, Three-Way Four-Position Ball Valve, 3/4" NPT(F) EW-98710-01 Mfr # 253851
COMPARE
14 0.75
$75.60USD / Each
053232 True Union, Three-Way Four-Position Ball Valve, 1-1/2" NPT(F) EW-98710-04 Mfr # 253854
COMPARE
56 1.5
$173.70USD / Each
VTBV108 True Union, Three-Way Four-Position Ball Valve, 2" NPT(F) EW-98710-05 Mfr # 253855
COMPARE
100 2
$247.70USD / Each