Tuff-Job Scrim-Reinforced Wipers

100% recycled material
For all of your cleaning needs, these disposable wipers are up to the task. Multiple plies offer excellent absorbency and the polyester webbing provides strength and durability. Superior wipe-dry capabilities make these low-lint wipers perfect for glass and other smooth surfaces. Also work well with grease and oil.
$7.00 - $94.50USD / of 1

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Format Description
Availability Pricing
Tuff-Job 626715-0001 Scrim Wipes 4ply, 9 3/4" x 16 3/4", Pop-Up, 150 Box EW-86115-97 Mfr # 626715-0001
COMPARE
Wipes Scrim Wipes 4ply, 9 3/4" x 16 3/4", Pop-Up, 150 Box
$21.50USD / of 1
Tuff-Job 626715-0002 Scrim Wipes 4ply, 13 1/4" x 12", Flat, 900 Box EW-86115-98 Mfr # 626715-0002
COMPARE
Wipes Scrim Wipes 4ply, 13 1/4" x 12", Flat, 900 Box
$94.50USD / of 1
Tuff-Job 626715-0003 Scrim Wipes 4ply, 12" x 13", Polybag, 50 EW-86115-99 Mfr # 626715-0003
COMPARE
Wipes Scrim Wipes 4ply, 12" x 13", Polybag, 50
$7.00USD / of 1
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat