Tunair No-Baffle Shake Flasks, 300 ml

Increase culture yields with optimum growth conditions for aerobic organisms
  • Turbo vane closure design increases oxygenation compared to standard Erlenmeyer flasks
  • Cap with removable filter lining protects contents of flask from airborne particulates and eliminates need to flame neck
  • All components are autoclavable
  • Suitable for culturing aerobic microbes, mammalian, and plant cells
These shake flasks work with any standard orbital shaker having adjustable speeds from 150 to 400 rpm. The unbaffled models behave similarly to Erlenmeyer flasks under these conditions.

Cleanup is easy—just soak in a mild dish soap to remove dirt, and air dry. All pieces are autoclavable, including the filter linings. Filter linings are also replaceable, with silicone membrane and Dri-Gauze materials available.

$40.00 - $540.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Cap Type Material Description
Availability Pricing
Tunair SS-2011 No-Baffle Shake Flask, 300 ml, 1 EA EW-01835-11 Mfr # SS-2011
COMPARE
Not included Polypropylene No-Baffle Shake Flask, 300 ml, 1 EA
$40.00USD / Each
Tunair SS-4001 No-Baffle Shake Flask, 300 ml, 4 pk EW-01835-13 Mfr # SS-4001
COMPARE
Not included Polypropylene No-Baffle Shake Flask, 300 ml, 4 pk
$116.00USD / Pkg of 4
Tunair SS-2001C No-Baffle Flask Kit (Dri-Gauze), 300 ml, 1 kit EW-01835-15 Mfr # SS-2001C
COMPARE
2-piece with Dri-Gauze membrane Polypropylene No-Baffle Flask Kit (Dri-Gauze), 300 ml, 1 kit
$397.00USD / Each
Tunair SS-2001S No-Baffle Flask Kit (Silicone), 300 ml, 1 kit EW-01835-17 Mfr # SS-2001S
COMPARE
2-piece with silicone membrane Polypropylene No-Baffle Flask Kit (Silicone), 300 ml, 1 kit
$540.00USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat