Bel-Art® Two- and Three-Way Stopcocks with Hose Barb Connections
Easily adjust flow
  • Constructed of durable PVDF
  • Suitable for use to a maximum temperature of 464°F (240°C) and maximum pressure of 5 psi
Chemical and heat resistant PVDF stopcocks allow you to stop flow quickly. Easily connect hose barb connectors to flexible tubing for quick and easy installation.
$6.80 - $186.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Inlet Port Orifice (mm) Outlet Port
Availability Pricing
COMPARE
1/2" to 5/16" Barb 2 1/2" to 5/16" Barb
$115.00USD / Each
COMPARE
1/2" to 5/16" Barb 4 1/2" to 5/16" Barb
$111.00USD / Each
COMPARE
1/2" to 5/16" Barb 2 1/2" to 5/16" Barb
$128.00USD / Each
COMPARE
1/4" to 5/16" Barb 4 1/4" to 5/16"
$107.00USD / Each
Bel-Art 30888-0006 PVDF Three-Way Stopcock, 6 mm hose barb, 4 mm orifice EW-06705-10 Mfr # 30888-0006
COMPARE
6 mm Barb 4 6 mm Barb
$145.00USD / Each
Bel-Art 30888-0008 PVDF Three-Way Stopcock, 8 mm hose barb, 6 mm orifice EW-06705-12 Mfr # 30888-0008
COMPARE
8 mm Barb 6 8 mm Barb
$168.00USD / Each
Bel-Art 30888-0010 PVDF Three-Way Stopcock, 10 mm hose barb, 8 mm orifice EW-06705-14 Mfr # 30888-0010
COMPARE
10 mm Barb 8 10 mm Barb
$186.00USD / Each
228265 Two-Way Stopcock, HDPE, 10 mm hose barb, 7 psi max
Always in Stock
EW-98261-10 Mfr # 228265
COMPARE
10 mm Barb 6.8 10 mm Barb
$6.80USD / Each
228275 Two-Way Stopcock, HDPE, 14 mm hose barb, 7 psi max EW-98261-20 Mfr # 228275
COMPARE
14 mm Barb 10.7 14 mm Barb
$6.80USD / Each
228285 Two-Way Stopcock, HDPE, 16 mm hose barb, 7 psi max EW-98261-30 Mfr # 228285
COMPARE
16 mm Barb 12.7 16 mm Barb
$6.80USD / Each
Showing 1- 10 of 10