Two-Way, Three-Way, and Sample Diaphragm Valves 48573
These diaphragm valves are made of PFA and feature no exposed metal parts. Choose between a two-way, three-way or sample diaphragm valve with various temperature and pressure ranges. Two-way valve 31101-05 includes bracket and screws. Three-way and sample valves include screws for mounting hardware.
$331.00 - $504.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Cv Inlet Port Outlet Port
Availability Pricing
QV2-144-M Diaphragm Valve, PFA, 1/4" NPT(F), 75 psi max EW-30502-06 Mfr # QV2-144-M
COMPARE
0.47 1/4" NPT(F) 1/4" NPT(F)
$458.00USD / Each
QV2-186-M Diaphragm Valve, PFA, 1/2" NPT(F), 75 psi max EW-30502-07 Mfr # QV2-186-M
COMPARE
1.17 1/2" NPT(F) 1/2" NPT(F)
$504.00USD / Each
Parker Hannifin MV-11-005 Two-Way Diaphragm Valve, 1/2" NPT(F), up to 100 psi EW-31101-03 Mfr # MV-11-005
COMPARE
3 1/2" NPT(F) 1/2" NPT(F)
$502.00USD / Each
MV-10-1044-02 Diaphragm Valve, PFA, 1/4" NPT(F), 29"Hg to 100 psi EW-31101-05 Mfr # MV-10-1044-02
COMPARE
0.47 1/4" NPT(F) 1/4" NPT(F)
$331.00USD / Each
Parker Hannifin MV-10-1344-00 Sample Diaphragm Valve, 1/4" NPT(F), up to 60 psi EW-31101-07 Mfr # MV-10-1344-00
COMPARE
0.55 to 1.10 1/4" NPT(F) 1/4" NPT(F)
$402.00USD / Each
COMPARE
0.9 1/4" NPT(F) 1/4" NPT(F)
$380.00USD / Each
COMPARE
0.95 or 0.9 1/4" NPT(F) 1/4" NPT(F)
$407.00USD / Each
MV-10-1344-02 Three-Way Diaphragm Valve, PFA, 1/4" NPT(F), 27"Hg to 45 psi EW-98403-08 Mfr # MV-10-1344-02
COMPARE
0.95 1/4" NPT(F) 1/4" NPT(F)
$399.00USD / Each
Showing 1- 8 of 8