Tygon 2475 I.B. High-Purity Reinforced Tubing

Formulation: TYGON® 2475 I.B.
Applications: Pharmaceutical and cosmetic processing, deionized water systems, cell harvest and media process systems, chemical transfer, preservative and sterile fill lines.
Characteristics: Plasticizer free, smooth inner surface (inhibits particulate build-up and bacterial growth), and nontoxic. Transparent.
Certification: Resin is FDA-compliant (21 CFR 177.1550) and meets NSF criteria. Tubing is manufactured according to GMP.
Temperature range: -94 to 125°F (-70 to 52°C)
Sterilize: With ethylene oxide or gamma irradiation.
$363.00 - $770.00USD / Pkg of 1

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
225 1/4 1/2
$363.00USD / Pkg of 1
COMPARE
210 3/8 5/8
$430.00USD / Pkg of 1
COMPARE
230 1/2 3/4
$610.00USD / Pkg of 1
COMPARE
135 5/8 7/8
$706.00USD / Pkg of 1
COMPARE
135 3/4 1 1/16
$770.00USD / Pkg of 1
COMPARE
125 1 1
$670.00USD / Pkg of 1
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat