Tygon® 3350 Platinum-Cured Silicone Tubing

Reduced particle entrapment with ultra-smooth inner surface
  • Minimal extractables help maintain fluid integrity
Formulation: TYGON® 3350 silicone, platinum cured.

Applications: Pharmaceutical, cosmetic, industrial, and auto-analysis applications.

Characteristics: Ultra-smooth inner surface reduces particle entrapment and minimizes bacterial growth. Good for mild to medium concentration bases, salts, and alcohols; is odorless, tasteless, and non-toxic. Transparent.

Certification: Elastomer is FDA-compliant (21 CFR 177.2600); meets USP Class VI requirements, NSF criteria and EP 3.2.9 requirements. Tubing is manufactured according to GMP.

Temperature range: –112 to 392°F (–80 to 200°C)

Sterilize: With ethylene oxide, gamma irradiation, or autoclave for 30 min, 15 psi.
$133.00 - $593.00USD / Pkg of 1

17 variations of this product are available.

See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
Tygon Transfer Tubing, BioPharm Platinum-Cured Silicone, 1/32" ID x 3/32" OD; 50 Ft
Always in Stock
EW-95702-00 Mfr # F00001
COMPARE
16 1/32 3/32
$133.00USD / Pkg of 1
Tygon Transfer Tubing, BioPharm Platinum-Cured Silicone, 1/16" ID x 1/8" OD; 50 Ft EW-95702-01 Mfr # F00002
COMPARE
10 1/16 1/8
$136.00USD / Pkg of 1
Tygon Transfer Tubing, BioPharm Platinum-Cured Silicone, 1/16" ID x 3/16" OD; 50 Ft EW-95702-02 Mfr # F00003
COMPARE
15 1/16 3/16
$179.00USD / Pkg of 1
Tygon Transfer Tubing, BioPharm Platinum-Cured Silicone, 3/32" ID x 5/32" OD; 50 Ft EW-95702-03 Mfr # F00004
COMPARE
7 3/32 5/32
$160.00USD / Pkg of 1
Tygon Transfer Tubing, BioPharm Platinum-Cured Silicone, 3/32" ID x 7/32" OD; 50 Ft EW-95702-04 Mfr # F00005
COMPARE
12 3/32 7/32
$212.00USD / Pkg of 1
Tygon Transfer Tubing, BioPharm Platinum-Cured Silicone, 1/8" ID x 3/16" OD; 50 Ft EW-95702-05 Mfr # F00006
COMPARE
7 1/8 3/16
$164.00USD / Pkg of 1
Tygon Transfer Tubing, BioPharm Platinum-Cured Silicone, 1/8" ID x 1/4" OD; 50 Ft
Always in Stock
EW-95702-06 Mfr # F00007
COMPARE
9 1/8 1/4
$221.00USD / Pkg of 1
Tygon Transfer Tubing, BioPharm Platinum-Cured Silicone, 3/16" ID x 1/4" OD; 50 Ft EW-95702-08 Mfr # F00009
COMPARE
5 3/16 1/4
$201.00USD / Pkg of 1
Tygon Transfer Tubing, BioPharm Platinum-Cured Silicone, 3/16" ID x 5/16" OD; 50 Ft EW-95702-09 Mfr # F00010
COMPARE
7 3/16 5/16
$277.00USD / Pkg of 1
Tygon Transfer Tubing, BioPharm Platinum-Cured Silicone, 3/16" ID x 3/8" OD; 50 Ft EW-95702-10 Mfr # F00011
COMPARE
9 3/16 3/8
$401.00USD / Pkg of 1
Tygon Transfer Tubing, BioPharm Platinum-Cured Silicone, 1/4" ID x 3/8" OD; 50 Ft EW-95702-13 Mfr # F00014
COMPARE
5 1/4 3/8
$296.00USD / Pkg of 1
Tygon Transfer Tubing, BioPharm Platinum-Cured Silicone, 1/4" ID x 7/16" OD; 50 Ft EW-95702-14 Mfr # F00015
COMPARE
7 1/4 7/16
$462.00USD / Pkg of 1
Tygon Transfer Tubing, BioPharm Platinum-Cured Silicone, 5/16" ID x 7/16" OD; 50 Ft EW-95702-16 Mfr # F00017
COMPARE
5 5/16 7/16
$414.00USD / Pkg of 1
Tygon Transfer Tubing, BioPharm Platinum-Cured Silicone, 5/16" ID x 1/2" OD; 50 Ft EW-95702-17 Mfr # F00018
COMPARE
6 5/16 1/2
$532.00USD / Pkg of 1
Tygon Transfer Tubing, BioPharm Platinum-Cured Silicone, 3/8" ID x 1/2" OD; 50 Ft EW-95702-18 Mfr # F00019
COMPARE
4 3/8 1/2
$451.00USD / Pkg of 1
Tygon Transfer Tubing, BioPharm Platinum-Cured Silicone, 3/8" ID x 9/16" OD; 50 Ft EW-95702-19 Mfr # F00020
COMPARE
5 3/8 9/16
$593.00USD / Pkg of 1
Tygon Transfer Tubing, BioPharm Platinum-Cured Silicone, 3/8" ID x 5/8" OD; 50 Ft EW-95702-20 Mfr # F00021
COMPARE
7 3/8 5/8
$443.00USD / Pkg of 1
Showing 1- 17 of 17

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat