Tygon® E-1000 Ultra-Soft Flexible Transfer Tubing

High degree of flexibility, performs well at low temperatures
  • Global market leader in range of formulations for most extensive chemical compatibility
Formulation: Tygon® E-1000
Applications: General laboratory, vent and drain lines. Ideal for lab installations requiring multiple directional changes, abrupt curves, or twisting.
Characteristics: High degree of flexibility, performs well at low temperatures, broad chemical resistance, resists kinking and collapse. Transparent.
Certification: FDA compliant. Made according to GMP.
Temperature range: –67 to 125°F (–55 to 52°C)
Sterilize:Autoclave or ethylene oxide.
$91.00 - $439.00USD / Pkg of 1

10 variations of this product are available.

See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
Tygon Transfer Tubing, Tygon® E-1000, 1/16" ID x 3/16" OD; 50 Ft
Buy More Save More
EW-65102-00 Mfr # ADK00003
COMPARE
26 1/16 3/16
$106.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-1000, 1/8" ID x 1/4" OD; 50 Ft
Buy More Save More
EW-65102-02 Mfr # ADK00007
COMPARE
16 1/8 1/4
$91.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-1000, 3/16" ID x 5/16" OD; 50 Ft
Buy More Save More
EW-65102-04 Mfr # ADK00012
COMPARE
10 3/16 5/16
$125.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-1000, 1/4" ID x 3/8" OD; 50 Ft
Buy More Save More
EW-65102-06 Mfr # ADK00017
COMPARE
8 1/4 3/8
$144.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-1000, 1/4" ID x 1/2" OD; 50 Ft
Buy More Save More
EW-65102-08 Mfr # ADK00019
COMPARE
14 1/4 1/2
$278.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-1000, 5/16" ID x 7/16" OD; 50 Ft
Buy More Save More
EW-65102-10 Mfr # ADK00022
COMPARE
7 5/16 7/16
$170.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-1000, 3/8" ID x 1/2" OD; 50 Ft
Buy More Save More
EW-65102-12 Mfr # ADK00027
COMPARE
7 3/8 1/2
$201.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-1000, 3/8" ID x 5/8" OD; 50 Ft
Buy More Save More
EW-65102-14 Mfr # ADK00029
COMPARE
11 3/8 5/8
$364.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-1000, 1/2" ID x 5/8" OD; 50 Ft
Buy More Save More
EW-65102-16 Mfr # ADK00036
COMPARE
6 1/2 5/8
$241.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-1000, 1/2" ID x 3/4" OD; 50 Ft
Buy More Save More
EW-65102-18 Mfr # ADK00038
COMPARE
8 1/2 3/4
$439.00USD / Pkg of 1
Showing 1- 10 of 10

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat