Tygon® E-Lab E-3603 Transfer Tubing

Long life, won’t age or oxidize
  • Global market leader in range of formulations for most extensive chemical compatibility
Formulation - E3603
Applications:General laboratory use.
Characteristics: Long life, won’t age or oxidize, able to bend to sharp radius curves, and easily works with fittings. Transparent.
Temperature range: –58 to 165°F (–50 to 74°C)
$33.10 - $921.00USD / Pkg of 1

29 variations of this product are available.

See All Product Options Below
29 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
Tygon Transfer Tubing, Tygon® E-Lab (E-3603), 1/32" ID x 3/32" OD; 50 Ft
Buy More Save More
EW-07407-70 Mfr # ACF00001
COMPARE
69 1/32 3/32
$33.10USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Lab (E-3603), 1/16" ID x 1/8" OD; 50 Ft
Buy More Save More
EW-07407-71 Mfr # ACF00002
COMPARE
36 1/16 1/8
$34.50USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Lab (E-3603), 1/16" ID x 3/16" OD; 50 Ft
Buy More Save More
EW-07407-72 Mfr # ACF00003
COMPARE
69 1/16 3/16
$87.40USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Lab (E-3603), 3/32" ID x 7/32" OD; 50 Ft
Buy More Save More
EW-07407-74 Mfr # ACF00005
COMPARE
47 3/32 7/32
$109.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Lab (E-3603), 1/8" ID x 3/16" OD; 50 Ft
Buy More Save More
EW-07407-75 Mfr # ACF00006
COMPARE
20 1/8 3/16
$65.60USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Lab (E-3603), 3/16" ID x 3/8" OD; 50 Ft
Buy More Save More
EW-07407-79 Mfr # ACF00013
COMPARE
36 3/16 3/8
$212.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Lab (E-3603), 1/4" ID x 3/8" OD; 50 Ft
Buy More Save More
EW-07407-80 Mfr # ACF00017
COMPARE
20 1/4 3/8
$284.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Lab (E-3603), 1/4" ID x 7/16" OD; 50 Ft
Buy More Save More
EW-07407-81 Mfr # ACF00018
COMPARE
28 1/4 7/16
$311.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Lab (E-3603), 5/16" ID x 7/16" OD; 50 Ft
Buy More Save More
EW-07407-83 Mfr # ACF00022
COMPARE
17 5/16 7/16
$204.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Lab (E-3603), 5/16" ID x 1/2" OD; 50 Ft
Buy More Save More
EW-07407-84 Mfr # ACF00023
COMPARE
23 5/16 1/2
$270.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Lab (E-3603), 3/8" ID x 1/2" OD; 50 Ft
Buy More Save More
EW-07407-86 Mfr # ACF00027
COMPARE
14 3/8 1/2
$219.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Lab (E-3603), 3/8" ID x 9/16" OD; 50 Ft
Buy More Save More
EW-07407-87 Mfr # ACF00028
COMPARE
20 3/8 9/16
$275.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Lab (E-3603), 3/8" ID x 5/8" OD; 50 Ft
Buy More Save More
EW-07407-88 Mfr # ACF00029
COMPARE
25 3/8 5/8
$434.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Lab (E-3603), 7/16" ID x 9/16" OD; 50 Ft
Buy More Save More
EW-07407-89 Mfr # ACF00032
COMPARE
13 7/16 9/16
$287.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Lab (E-3603), 1/2" ID x 5/8" OD; 50 Ft
Buy More Save More
EW-07407-90 Mfr # ACF00036
COMPARE
12 1/2 5/8
$254.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Lab (E-3603), 1/2" ID x 11/16" OD; 50 Ft
Buy More Save More
EW-07407-91 Mfr # ACF00037
COMPARE
15 1/2 11/16
$334.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Lab (E-3603), 1/2" ID x 3/4" OD; 50 Ft
Buy More Save More
EW-07407-92 Mfr # ACF00038
COMPARE
20 1/2 3/4
$522.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Lab (E-3603), 1/2" ID x 13/16" OD; 50 Ft
Buy More Save More
EW-07407-93 Mfr # ACF00039
COMPARE
24 1/2 13/16
$574.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Lab (E-3603), 5/8" ID x 7/8" OD; 50 Ft
Buy More Save More
EW-07407-95 Mfr # ACF00046
COMPARE
17 5/8 7/8
$528.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Lab (E-3603), 3/4" ID x 1-1/8" OD; 50 Ft
Buy More Save More
EW-07407-97 Mfr # ACF00055
COMPARE
20 3/4 1
$921.00USD / Pkg of 1
Showing 1 - 20 of 29

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat