Tygon High-Purity Tubing

Formulation: TYGON 2475
Applications: Sensitive fluid transfer applications requiring high purity.
Characteristics: Plasticizer free, smooth inner surface (inhibits particulate build-up and bacterial growth), safely dispose of through incineration, and nontoxic. Transparent.
Certification: Resin is FDA compliant and meets USP 23 Class VI requirements. Tubing is manufactured according to GMP.
Temperature range: -94 to 130°F (-70 to 54°C)
Sterilize: With ethylene oxide or gamma irradiation.
$116.00 - $660.00USD / Pkg of 1

10 variations of this product are available.

See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
85 1/16 3/16
$116.00USD / Pkg of 1
COMPARE
50 1/8 1/4
$172.00USD / Pkg of 1
COMPARE
40 3/16 5/16
$189.00USD / Pkg of 1
COMPARE
30 1/4 3/8
$203.00USD / Pkg of 1
COMPARE
18 5/16 7/16
$236.00USD / Pkg of 1
COMPARE
20 3/8 1/2
$246.00USD / Pkg of 1
COMPARE
29 1/2 3/4
$528.00USD / Pkg of 1
COMPARE
25 5/8 7/8
$622.00USD / Pkg of 1
COMPARE
21 3/4 1
$660.00USD / Pkg of 1
Tygon High Purity Tubing, 1"ID x 1-3/8"OD, 25 ft/pk
Buy More Save More
EW-95635-59 Mfr # ACG42064
COMPARE
20 1 1
$584.00USD / Pkg of 1
Showing 1- 10 of 10

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat