Tygon® S3™ E-Food B-44-4X Food, Milk, and Dairy Transfer Tubing

FDA compliant for food contact
  • Global market leader in range of formulations for most extensive chemical compatibility
Formulation: Tygon® S3-B-44-4X
Applications: Food and beverage processing and dispensing, milk and dairy processing.
Characteristics: Bio-based, phthalate-free flexible. Nonwetting, odorless, tasteless, and nontoxic. Transparent.
Certification: Resin is FDA compliant, NSF-listed (Standard 51), and meets 3A standards (20-27) for food and beverage handling.
Temperature range: –49 to 165°F (–45 to 74°C)
Sterilize: Autoclave for 30 min at 250°F, 15 psi or ethylene oxide.
$47.75 - $553.00USD / Pkg of 1

10 variations of this product are available.

See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
Tygon Transfer Tubing, Tygon® E-Food (B-44-4X), 1/16" ID x 3/16" OD; 50 Ft
Buy More Save More
EW-95644-01 Mfr # AAA00003
COMPARE
108 1/16 3/16
$47.75USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Food (B-44-4X), 1/8" ID x 1/4" OD; 50 Ft
Buy More Save More
EW-95644-03 Mfr # AAA00007
COMPARE
60 1/8 1/4
$67.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Food (B-44-4X), 3/16" ID x 5/16" OD; 50 Ft
Buy More Save More
EW-95644-05 Mfr # AAA00012
COMPARE
43 3/16 5/16
$89.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Food (B-44-4X), 1/4" ID x 3/8" OD; 50 Ft
Buy More Save More
EW-95644-07 Mfr # AAA00017
COMPARE
34 1/4 3/8
$114.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Food (B-44-4X), 5/16" ID x 7/16" OD; 50 Ft
Buy More Save More
EW-95644-09 Mfr # AAA00022
COMPARE
28 5/16 7/16
$133.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Food (B-44-4X), 3/8" ID x 1/2" OD; 50 Ft
Buy More Save More
EW-95644-11 Mfr # AAA00027
COMPARE
25 3/8 1/2
$157.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Food (B-44-4X), 1/2" ID x 3/4" OD; 50 Ft
Buy More Save More
EW-95644-13 Mfr # AAA00038
COMPARE
34 1/2 3/4
$334.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Food (B-44-4X), 5/8" ID x 7/8" OD; 50 Ft
Buy More Save More
EW-95644-15 Mfr # AAA00046
COMPARE
29 5/8 7/8
$389.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Food (B-44-4X), 3/4" ID x 1" OD; 50 Ft
Buy More Save More
EW-95644-17 Mfr # AAA00053
COMPARE
25 3/4 1
$443.00USD / Pkg of 1
Tygon Transfer Tubing, Tygon® E-Food (B-44-4X), 1" ID x 1-1/4" OD; 50 Ft
Buy More Save More
EW-95644-19 Mfr # AAA00062
COMPARE
20 1 1 1/4
$553.00USD / Pkg of 1
Showing 1- 10 of 10

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat