UL-Listed Air-Operated Double Diaphragm Pump
Designed to meet UL79 Standards for handling flammable liquids
  • Fully groundable to prevent static discharge
  • Handles abrasive and shear-sensitive materials
  • Self-priming up to 24 ft of water
  • Deadhead against closed discharge without damage to the pump
$1,289.50 - $1,696.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Air Inlet Pressure (PSI) Max Flow Rate (GPM) Max Input VDC (VDC)
Availability Pricing
Warren Rupp U1FB1XBTXNS600 Air-operated Diaphragm Pump, UL Listed, 1", 0-45gpm, Aluminium and PTFE construction EW-74440-90 Mfr # U1FB1XBTXNS600
COMPARE
100 45
$1,289.50USD / Each
Warren Rupp U1FB1XGTXNS600 Air-operated Diaphragm Pump, UL Listed, 1", 0-45gpm, Aluminium and Buna N EW-74440-95 Mfr # U1FB1XGTXNS600
COMPARE
100 45
$1,696.50USD / Each