Ultrapure Application Filter Cartridges
These cartridges are autoclavable to 250°F <121°C) and in-line sterilizable to 257°F (125°C) for up to 30-minute cycles. They are tolerant to an accumulated exposure of 10 hours. The filters may also be sanitized with compatible chemical agents. All cartridges are suitable to 180°F (82°C) at 10 psid.
The cartridges comply with U.S. CFR Title 21 guidelines for repeated food contact, comply with USP Class VI, 121°C Plastics guidelines, and pass the MEM Elution Cytotoxicity Test. Aqueous extracts contain less than 0.25 EU/mL.
$87.50 - $404.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Length (in) Filter Media Micron Rating
Availability Pricing
COMPARE
10 Nylon 0.22
$236.00USD / Each
COMPARE
10 Nylon 0.45
$217.00USD / Each
COMPARE
10 Glass fiber 3
$124.00USD / Each
COMPARE
10 Polypropylene 1
$121.00USD / Each
COMPARE
10 Polypropylene 3
$114.00USD / Each
COMPARE
10 Polypropylene 5
$112.00USD / Each
COMPARE
10 Polypropylene 10
$87.50USD / Each
COMPARE
10 PTFE 0.1
$404.00USD / Each
COMPARE
10 PTFE 0.2
$399.00USD / Each
COMPARE
10 Polyethersulfone (PES) 0.2
$220.00USD / Each
Showing 1- 10 of 10