Union Fittings, Female Threaded, Straight, PTFE
Chemically-inert PTFE handles most aggressive chemicals
  • Constructed of durable PTFE
Chemically inert, PTFE offers outstanding resistance for any use, especially for critical high-purity applications. When subjected to higher pressure, PTFE as a material has a tendency to creep over time. As a result, these fittings work best at lower operating pressures (110 psi maximum operating pressure at ambient temperature for smaller sizes). All fittings are made from virgin PTFE material.
$44.25 - $135.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Thread size NPT(F) (port 1) (in) Thread size NPT(F) (port 2) (in)
Availability Pricing
COMPARE
PTFE 1/8 1/8
$44.25USD / Each
COMPARE
PTFE 3/8 3/8
$64.50USD / Each
COMPARE
PTFE 1/2 1/2
$84.00USD / Each
COMPARE
PTFE 3/4 3/4
$114.00USD / Each
COMPARE
PTFE 1 1
$135.00USD / Each