Uvex by Honeywell Bionic Face Shield
Available for sale in the US only
  • Full-shield design provides built-in chin and top-of-head protection
  • Lightweight design, foam backing, and customizable fit make this face shield ideal for extended use
  • Fits comfortably over most goggles and respirators
  • Compliance: ANSI Z87.1-2003, CSA Z94.3
Shield provides secondary protection for workers exposed to falling or flying objects, impact, chemical splash, and airborne debris. Full shield design provides built-in chin and top-of-head protection. Lightweight design, foam backing, and customizable fit make this face shield ideal for extended use. Fits comfortably over most goggles and respirators.
Compliance: ANSI Z87.1-2003 and CSA Z94.3
$10.70 - $60.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Lens Coating Lens Color
Availability Pricing
Uvex by Honeywell Bionic Face Shield, Clear, uncoated
Buy More Save More
EW-81690-11 Mfr # S8500
COMPARE
Uncoated Clear
$41.75USD / Each
Uvex by Honeywell Bionic Face Shield, Clear, Antifog coating
Buy More Save More
EW-81690-12 Mfr # S8510
COMPARE
Antifog Clear
$60.00USD / Each
Uvex by Honeywell Bionic Replacement Visor, Uncoated Polycarbonate
Buy More Save More
EW-81690-13 Mfr # S8550
COMPARE
Uncoated Clear
$10.70USD / Each
Uvex by Honeywell Bionic Replacement Visor, Clear, antifog hardcoat
Buy More Save More
EW-81690-14 Mfr # S8555
COMPARE
Antifog Clear
$34.25USD / Each