Uvex by Honeywell Genesis Safety Glasses
Available for sale in the US only
  • Sleek, stylish design for cool, comfortable protection
  • Lenses exceed, by seven times, the standards set by ANSI for impact velocity
Wraparound lens design provides uncompromised peripheral vision. Internal points of contact serve as a shock absorber to diffuse impact energy away from eyes.
A ventilation channel between the frame and lens allows moisture to escape — preventing fogging. Coating helps to keep your lenses scratch free. Lenses and temple stems are fully adjustable to provide a custom fit. Compliance: ANSI Z87.1-2010, CSA Z94.3, military V0
$19.40 - $20.20USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Frame Color Lens Color Compliance
Availability Pricing
Uvex by Honeywell S3200 Black frame glasses with clear lenses
Buy More Save More
EW-86325-70 Mfr # S3200
COMPARE
Black Clear ANSI Z87.1-2010; CSA Z94.3; military V0
$20.20USD / Each
Uvex by Honeywell S3240 Translucent blue frame glasses with clear lenses
Buy More Save More
EW-86325-80 Mfr # S3240
COMPARE
Blue Clear ANSI Z87+ (High Impact); CSA Z94.3; military V0
$19.40USD / Each