Uvex by Honeywell Seismic Safety Eyewear
Available for sale in the US only
  • Range of comfort and adjustability features for all-day wear
Flexible design allows you to wear these as safety glasses, or attach the flame-resistant headband to wear as goggles. Also, leave the face cushioning inner frame on for extra comfort during extended-wear applications, remove it for a standard safety glasses feel. The cushioning seals your eyes from dirty, high-particulate work environments such as grinding or sanding. Wraparound style enhances peripheral vision and eye coverage. Top and bottom venting helps minimize fogging.
Compliance: ANSI Z87.1-2010 (with or without cushioning), CSA Z94.3 (with cushioning only)
$27.60 - $36.60USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Lens Coating Lens Color
Availability Pricing
Uvex by Honeywell S0600 Safety Eyewear, Clear lens
Buy More Save More
EW-86497-60 Mfr # S0600
COMPARE
Ultra-dura® (hard coating) Clear
$27.60USD / Each
Uvex by Honeywell S0600X Safety Eyewear, Clear lens, Antifog coating
Buy More Save More
EW-86497-61 Mfr # S0600X
COMPARE
Antifog Clear
$30.90USD / Each
Uvex by Honeywell S0600D Safety Eyewear, Clear lens, Anti-Fog, Dual Hard-Coat
Buy More Save More
EW-86497-62 Mfr # S0600D
COMPARE
Antifog/Hard Coat Clear
$36.60USD / Each