V-Watch Personal Voltage Detector
Always on for added protection
  • Audible (100 dB) and visual alarms go off in the presence of electrical fields or energized electrical equipment
  • Typical warning distance of 7 feet (2.1 meters) from a 4 kVAC conductor—distance increases with higher voltages
  • Handy lanyard case lets you wear the detector around your neck at mid-torso level—no need to carry anything extra
  • Low-battery indicator and field replaceable batteries—accepts one 9 V battery
The V-Watch personal voltage detector offers an additional line of protection for line crews, storm restoration workers, and first responders against AC voltages (50/60 Hz sensing threshold). The audible (100 dB) and visual alarms go off more frequently as the wearer gets closer to energized equipment. Rugged housing is ideal for outdoor use and protects the detector from falling rain.
Included lanyard carrying case is length adjustable—ideal for wearing at the optimal mid-torso level. Plus the case is made with a flame-resistant material that is a highly visible, safety-green color. The detector does not work inside the closed lanyard case; open the case when in use, otherwise close to conserve battery life.
$505.75 - $549.25USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy True Rms
Availability Pricing
HD Electric VW-20HFR Personal Voltage Detector with Lanyard EW-23022-60 Mfr # VW-20HFR
COMPARE
$505.75USD / Each
HD Electric VW-20HFR-K Personal Voltage Detector Kit with Lanyard & Case EW-23022-65 Mfr # VW-20HFR-K
COMPARE
$549.25USD / Each