VACUUBRAND® Compact Diaphram Vacuum Pumps for Filtration
High performance at an affordable price
  • Excellent for filtration and other pressure differential applications
  • Quiet and ultra-low maintenance
These vacuum pumps deliver high performance in a compact, space saving design. Fluoropolymer valves and diaphragms provide solvent resistance. Choose Chemistry model for acidic vapors, or the aluminum head version for economy. Typical service interval is in excess of 15,000 operating hours provides for virtually maintanence free operation.
$990.00 - $1,377.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Vacuum (torr) Wetted Parts Description
Availability Pricing
Vacuubrand ME 1 Standard Duty Vacuum Pump, 0.5 cfm, 75 Torr; 120V EW-79104-01 Mfr # 20721003
COMPARE
76.7 PTFE, Aluminum, Polyamide, Polyethylene ME1 Vacuum Pump, 0.5 CFM, 75 Torr, 120 VAC, 50/60 Hz
$990.00USD / Each
Vacuubrand ME 1C Chemistry Design Vacuum Pump, 0.5 cfm, 75 Torr; 120V EW-79104-03 Mfr # 20721103
COMPARE
76.7 PTFE, ETFE Vacuum Pump, 0.5 CFM, Chemical Duty, 120 VAC, 50/60 Hz
$1,377.00USD / Each
Vacuubrand ME 1C Chemistry Design Vacuum Pump, 0.5 cfm, 75 Torr; 230V EW-79104-04 Mfr # 20721100
COMPARE
76.7 PTFE, ETFE Vacuum Pump, 0.5 CFM, Chemical Duty, 230 VAC, 50/60 Hz
$1,377.00USD / Each