Vaisala CO2 Transmitters for Harsh Environments
These transmitters are designed to measure CO2 in harsh and humid environments. The housing is dust and waterproof to NEMA 4/IP65 standards and materials are corrosion resistant.
The transmitters are well suited for demanding industrial CO2 applications, such as:
  • Horticulture and fruit storage
  • Greenhouses and mushroom farming
  • Safety alarming and leakage monitoring
  • Demand control ventilation in harsh environments
$1,448.00 - $1,498.50USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max CO2 (ppm) Max Humidity (RH) Max Oxygen (%)
Availability Pricing
Vaisala CO2 0/5% Transmitter Wall Mount with LED Lights EW-86493-08 Mfr # GMP251 C2A0A0N1 + INDIGO201 2A0A0
COMPARE
$1,498.50USD / Each
Vaisala CO2 0/20% Transmitter Wall Mount with LED Lights EW-86493-09 Mfr # GMP251 A2A0A0N1 + INDIGO201 2A0A0
COMPARE
$1,498.50USD / Each
Vaisala CO2 10K ppm Transmitter Wall Mount with LED Lights EW-86493-11 Mfr # GMP252 12A0A0N1 + INDIGO201 2A0A0
COMPARE
$1,448.00USD / Each