Vaisala Digital Barometers with/without Display
Ideal for demanding weather measurement applications
  • Displays World Meteorological Organization (WMO) pressure tendency codes
  • Simultaneously indicates barometric pressure, three hour pressure trend and WMO tendency code
Connect 1/8” inner diameter (ID) tubing to the barbed pressure fitting for barometric pressure measurements. The backlit display allows easy visibility in low light areas. Along with the barometric pressure, the transmitter will display the pressure tendency code and three-hour trend. Selectable analog output sends measurements to your other recording instruments.
$3,165.19 - $3,513.69USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description Barometric Pressure Accuracy (PSI)
Availability Pricing
Vaisala Digital Barometer, without Display EW-99790-00 Mfr # PTB330B0ACJAACEB0A0B
COMPARE
Digital Barometer, without display ±0.007\" Hg (0.25 mbar)
$3,165.19USD / Each
Vaisala Digital Barometer, with Display EW-99790-02 Mfr # PTB330B1ACJAACEB0A0B
COMPARE
Digital Barometer, with display ±0.007\" Hg (0.25 mbar)
$3,513.69USD / Each