Value Plastics ADCF Polypropylene Sanitary Adapters
No BSE documentation required!
These adapters are perfect to transition from sanitary clamp connections to a barb for flexible tubing. All adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow and no particulate build-up. There are no sharp edges in the flow path, so cleaning is very easy. Made from animal-derived ingredient-free polypropylene, these adapters offer good chemical resistance and may be sterilized by gamma irradiation. Material meets the requirements for USP Class VI. Available with 3/8'' or 1/2'' hose barbs and 3/4'' mini or 1-1/2'' standard sanitary clamp connections.
$22.25 - $36.50USD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Sanitary Clamp Size (Port 1) (in) Description
Availability Pricing
COMPARE
3/4 Barbed to Sanitary Adapter, 3/8" ID to Mini 3/4" ADCF-PP
$26.75USD / Each
Value Plastics Barbed to Sanitary Adapter, ADCF Polypropylene; 1/2" ID to Mini 3/4" EW-31513-14 Mfr # SFMN680-6005
COMPARE
3/4 Barbed to Sanitary Adapter, 1/2" ID to Mini 3/4" ADCF-PP
$25.75USD / Each
Value Plastics Barbed to Sanitary Adapter, ADCF Polypropylene; 3/8" ID to 1-1/2"
Always in Stock
EW-31513-15 Mfr # SFMX670-6005
COMPARE
1-1/2 Barbed to Sanitary Adapter, 3/8" ID to 1-1/2", ADCF-PP
$30.25USD / Each
Value Plastics Barbed to Sanitary Adapter, ADCF Polypropylene; 1/2" ID to 1-1/2"
Always in Stock
EW-31513-16 Mfr # SFMX680-6005
COMPARE
1-1/2 Barbed to Sanitary Adapter, 1/2" ID to 1-1/2", ADCF-PP
$26.25USD / Each
Value Plastics Sanitary End Cap, ADCF Polypropylene; Mini 3/4" EW-31513-17 Mfr # SFMNC-6005
COMPARE
3/4 Sanitary End Cap, Mini 3/4", ADCF-PP
$22.25USD / Each
Value Plastics Sanitary End Cap, ADCF Polypropylene; 1-1/2"
Always in Stock
EW-31513-18 Mfr # SFMXC-6005
COMPARE
1-1/2 Sanitary End Cap, 1-1/2", ADCF-PP
$28.25USD / Each
Value Plastics Barbed to Sanitary Adapter, ADCF Polypropylene; 1/4" ID to Mini 3/4" EW-31513-51 Mfr # SFMN655-6005
COMPARE
1/2 Barbed to Sanitary Adapter, 1/4" ID to Mini 3/4" ADCF-PP
$24.25USD / Each
Value Plastics Barbed to Sanitary Adapter, ADCF Polypropylene; 3/4" ID to Mini 3/4" EW-31513-54 Mfr # SFMN6100-6005
COMPARE
3/4 Barbed to Sanitary Adapter, 3/4" ID to Mini 3/4" ADCF-PP
$33.50USD / Each
Value Plastics Barbed to Sanitary Adapter, ADCF Polypropylene; 1/4" ID to 1-1/2"
Always in Stock
EW-31513-55 Mfr # SFMX655-6005
COMPARE
1-1/2 Barbed to Sanitary Adapter, 1/4" ID to 1-1/2", ADCF-PP
$28.75USD / Each
Value Plastics Barbed to Sanitary Adapter, ADCF Polypropylene; 3/4" ID to 1-1/2" EW-31513-58 Mfr # SFMX6100-6005
COMPARE
1-1/2 Barbed to Sanitary Adapter, 3/4" ID to 1-1/2", ADCF-PP
$36.00USD / Each
Value Plastics Barbed to Sanitary Adapter, ADCF Polypropylene; 1" ID to 1-1/2" EW-31513-59 Mfr # SFMX6110-6005
COMPARE
1-1/2 Barbed to Sanitary Adapter, 1" ID to 1-1/2", ADCF-PP
$36.50USD / Each
Showing 1- 11 of 11