Value Plastics Certified Sanitary Adapters, ADCF Polypropylene
Certificate of compliance included!
  • Animal derivative component free
  • Meet USP Class IV requirements
  • Sterilizable vis gamma irradiation
These sanitary fittings are the perfect to transition from sanitary clamp connections to seal or barb for flexible tubing. All adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow—and no particulate build-up. There are no sharp edges in the flow path, so cleaning is very easy. Made from animal-derived ingredient-free polypropylene, these adapters offer good chemical resistance and may be sterilized by gamma irradiation. Material meets the requirements for USP Class VI.
$210.00 - $303.00USD / Pkg of 10
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connection Type Sanitary Clamp Size (Port 1) (in) Description
Availability Pricing
Value Plastics Barbed to Sanitary Adapter, 3/8" ID to 3/4" Mini, ADCF PP; 10/Pk EW-31515-89 Mfr # SFMN670-6005
COMPARE
Sanitary x Barb 3/4 Barbed to Sanitary Adapter, 3/8" ID to 3/4" Mini, ADCF PP; 10/Pk
$218.00USD / Pkg of 10
Value Plastics Barbed to Sanitary Adapter, 1/2" ID to 3/4" Mini, ADCF PP; 10/Pk EW-31515-90 Mfr # SFMN680-6005
COMPARE
Sanitary x Barb 3/4 Barbed to Sanitary Adapter, 1/2" ID to 3/4" Mini, ADCF PP; 10/Pk
$223.00USD / Pkg of 10
Value Plastics Barbed to Sanitary Adapter, 3/8" ID to 1-1/2", ADCF PP; 10/Pk EW-31515-91 Mfr # SFMX670-6005
COMPARE
Sanitary x Barb 1-1/2 Barbed to Sanitary Adapter, 3/8" ID to 1-1/2", ADCF PP; 10/Pk
$223.00USD / Pkg of 10
Value Plastics Barbed to Sanitary Adapter, 1/2" ID to 1-1/2", ADCF PP; 10/Pk EW-31515-92 Mfr # SFMX680-6005
COMPARE
Sanitary x Barb 1-1/2 Barbed to Sanitary Adapter, 1/2" ID to 1-1/2", ADCF PP; 10/Pk
$253.00USD / Pkg of 10
Value Plastics Sanitary End Cap, 3/4" Mini, ADCF PP; 10/Pk EW-31515-93 Mfr # SFMNC-6005
COMPARE
Sanitary x End cap 3/4 Sanitary End Cap, 3/4" Mini, ADCF PP; 10/Pk
$210.00USD / Pkg of 10
Value Plastics Sanitary End Cap, 1-1/2", ADCF PP; 10/Pk EW-31515-94 Mfr # SFMXC-6005
COMPARE
Sanitary x End cap 1-1/2 Sanitary End Cap, 1-1/2", ADCF PP; 10/Pk
$210.00USD / Pkg of 10
Value Plastics Barbed to Sanitary Adapter, 1/4" ID to 3/4" Mini, ADCF PP; 10/Pk EW-31515-95 Mfr # SFMN655-6005
COMPARE
Sanitary x Barb 3/4 Barbed to Sanitary Adapter, 1/4" ID to 3/4" Mini, ADCF PP; 10/Pk
$216.00USD / Pkg of 10
Value Plastics Barbed to Sanitary Adapter, 3/4" ID to 3/4" Mini, ADCF PP; 10/Pk EW-31515-96 Mfr # SFMN6100-6005
COMPARE
Sanitary x Barb 3/4 Barbed to Sanitary Adapter, 3/4" ID to 3/4" Mini, ADCF PP; 10/Pk
$251.00USD / Pkg of 10
Value Plastics Barbed to Sanitary Adapter, 1/4" ID to 1-1/2", ADCF PP; 10/Pk EW-31515-97 Mfr # SFMX655-6005
COMPARE
Sanitary x Barb 1-1/2 Barbed to Sanitary Adapter, 1/4" ID to 1-1/2", ADCF PP; 10/Pk
$220.00USD / Pkg of 10
Value Plastics Barbed to Sanitary Adapter, 3/4" ID to 1-1/2", ADCF PP; 10/Pk EW-31515-98 Mfr # SFMX6100-6005
COMPARE
Sanitary x Barb 1-1/2 Barbed to Sanitary Adapter, 3/4" ID to 1-1/2", ADCF PP; 10/Pk
$289.00USD / Pkg of 10
Value Plastics Barbed to Sanitary Adapter, 1" ID to 1-1/2", ADCF PP; 10/Pk EW-31515-99 Mfr # SFMX6110-6005
COMPARE
Sanitary x Barb 1-1/2 Barbed to Sanitary Adapter, 1" ID to 1-1/2", ADCF PP; 10/Pk
$303.00USD / Pkg of 10
Showing 1- 11 of 11