Value Plastics PVDF Sanitary Adapters
These adapters are perfect to transition from sanitary clamp connections to a barb for flexible tubing. All adapters feature a flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow without particulate build-up. There are no sharp edges in the flow path to make cleaning very easy. These adapters offer good chemical resistance and may be sterilized by gamma irradiation, EtOH, or autoclave. Material meets the requirements for USP Class VI, ADCF, FDA, and are BPA-free.
$31.50 - $44.50USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Sanitary Clamp Size (Port 1) (in) Description
Availability Pricing
Value Plastics Barbed to sanitary adapter, 1/4" to mini sanitary, PVDF EW-31513-61 Mfr # SFMN655-J1A
COMPARE
1/4 1/2" Mini Barbed to sanitary adapter, 1/4" to mini sanitary, PVDF
$37.00USD / Each
Value Plastics Barbed to sanitary adapter, 3/8" to mini sanitary, PVDF
Always in Stock
EW-31513-62 Mfr # SFMN670-J1A
COMPARE
3/8 1/2" Mini Barbed to sanitary adapter, 3/8" to mini sanitary, PVDF
$35.50USD / Each
Value Plastics Barbed to sanitary adapter, 1/2" to mini sanitary, PVDF EW-31513-63 Mfr # SFMN680-J1A
COMPARE
1/2 1/2" Mini Barbed to sanitary adapter, 1/2" to mini sanitary, PVDF
$43.25USD / Each
Value Plastics Barbed to sanitary adapter, 3/4" to mini sanitary, PVDF EW-31513-64 Mfr # SFMN6100-J1A
COMPARE
3/4 1/2" Mini Barbed to sanitary adapter, 3/4" to mini sanitary, PVDF
$44.50USD / Each
Value Plastics Barbed to sanitary adapter, 1/4" to 1-1/2" sanitary, PVDF
Always in Stock
EW-31513-65 Mfr # SFMX655-J1A
COMPARE
1/4 1-1/2 Barbed to sanitary adapter, 1/4" to 1-1/2" sanitary, PVDF
$41.75USD / Each
Value Plastics Barbed to sanitary adapter, 3/8" to 1-1/2" sanitary, PVDF
Always in Stock
EW-31513-66 Mfr # SFMX670-J1A
COMPARE
3/8 1-1/2 Barbed to sanitary adapter, 3/8" to 1-1/2" sanitary, PVDF
$38.50USD / Each
Value Plastics Barbed to sanitary adapter, 1/2" to 1-1/2" sanitary, PVDF EW-31513-67 Mfr # SFMX680-J1A
COMPARE
1/2 1-1/2 Barbed to sanitary adapter, 1/2" to 1-1/2" sanitary, PVDF
$31.50USD / Each
Value Plastics Barbed to sanitary adapter, 3/4" to 1-1/2" sanitary, PVDF EW-31513-68 Mfr # SFMX6100-J1A
COMPARE
3/4 1-1/2 Barbed to sanitary adapter, 3/4" to 1-1/2" sanitary, PVDF
$38.50USD / Each
Value Plastics Barbed to sanitary adapter, 1" to 1-1/2" sanitary, PVDF EW-31513-69 Mfr # SFMX6110-J1A
COMPARE
1 1-1/2 Barbed to sanitary adapter, 1" to 1-1/2" sanitary, PVDF
$43.25USD / Each
Showing 1- 9 of 9