Variable-Speed Drives for Micropump Low-Flow Pump Heads
  • Compatible with Micropump® low-flow C-mount pump heads without canisters (order separately)
  • Magnetic seal minimizes friction—all horsepower is transmitted directly to the motor gears
  • Hundreds of hours of dependable, leakproof service
  • Variable speed operation with optional power supply/speed controller—order at side.
Order drives at side; order power supply/speed controller separately at side. Power supply includes a 2-amp fuse and a 6-ft, UL-listed, three-wire cord with plug for 115 VAC operation and measure 7 3/4"L x 5 1/2"W x 4 3/16"H.
$399.70 - $618.25USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Motor RPM (rpm) Power (amps) Power (VDC)
Availability Pricing
Micropump Variable speed drive for low-flow pump heads, 6000 rpm, 0 to 12 VDC EW-07002-37 Mfr # 83472 (MODEL #331A)
COMPARE
6000 2.5 0 to 12
$399.70USD / Each
Micropump 40045 Variable speed drive for low-flow pump heads, 8000 rpm, 0 to 24 VDC EW-07002-38 Mfr # 40045
COMPARE
8000 0.5 0 to 24
$477.80USD / Each
Micropump 40055 Variable speed drive for low-flow pump heads, 500 rpm, 0 to 24 VDC EW-07002-39 Mfr # 40055
COMPARE
500 0.6 0 to 24
$618.25USD / Each