Vector Laboratories Histology Slide Preparation Products

Prepare the best samples possible
Offering high sensitivity, low background, reliability, and reproducibility, these laboratory immunohistochemistry (IHC) slide preparation products accommodate a wide range of protocols.
$121.25 - $258.75USD / Each

12 variations of this product are available.

See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Vector Laboratories Vectabond Tissue Section Adhesive, 7 ml EW-93951-66 Mfr # SP-1800-7
COMPARE
Tissue Section Adhesive, 7 ml
$178.75USD / Each
Vector Laboratories ImmPRINT™ Solvent-Resistant Histology Pen, 5/pk EW-93951-67 Mfr # H-6100
COMPARE
Solvent-Resistant Histology Pen, 5/pk
$121.25USD / Pkg of 5
Vector Laboratories ImmEDGE™ Hydrophobic Barrier Pen for Immunohistochemistry and in situ Hybridization, 2/pk EW-93951-68 Mfr # H-4000
COMPARE
Hydrophobic Barrier Pen for Immunohistochemistry and in situ Hybridization, 2/pk
$220.00USD / Pkg of 2
Vector Laboratories H-3300 Citrate-Based Antigen Unmasking Solution, pH 6; 250 mL EW-93951-69 Mfr # H-3300-250
COMPARE
Citrate-Based Antigen Unmasking Solution, pH 6; 250 mL
$216.25USD / Each
Vector Laboratories Tris-based Antigen Unmasking Solution, pH 9; 250 mL EW-93951-70 Mfr # H-3301-250
COMPARE
Tris-based Antigen Unmasking Solution, pH 9, 25 L
$212.50USD / Each
Vector Laboratories Bloxall™ Dual-Enzyme Blocking Solution, 100 ml EW-93951-71 Mfr # SP-6000-100
COMPARE
Dual-Enzyme Blocking Solution, 100 ml
$171.25USD / Each
Vector Laboratories VECTASHIELD Antifade Mounting Medium, 10 ml EW-93952-23 Mfr # H-1000-10
COMPARE
$226.25USD / Each
Vector Laboratories VECTASHIELD Antifade Mounting Medium with DAPI, 10 ml EW-93952-24 Mfr # H-1200-10
COMPARE
$242.50USD / Each
Vector Laboratories VECTASHIELD® Antifade Mounting Medium with PI, 10 ml EW-93952-25 Mfr # H-1300-10
COMPARE
Antifade Mounting Medium with PI, 10 ml
$246.25USD / Each
Vector Laboratories VECTASHIELD® Hardset Antifade Mounting Medium, 10 ml EW-93952-26 Mfr # H-1400-10
COMPARE
Hardset Antifade Mounting Medium, 10 ml
$242.50USD / Each
Vector Laboratories VECTASHIELD® Hardset Antifade Mounting Medium with DAPI, 10 ml EW-93952-27 Mfr # H-1500-10
COMPARE
Hardset Antifade Mounting Medium with DAPI, 10 ml
$258.75USD / Each
Vector Laboratories VECTASHIELD® Hardset Antifade Mounting Medium with Phalloidin, 10 ml EW-93952-28 Mfr # H-1600-10
COMPARE
Hardset Antifade Mounting Medium with Phalloidin, 10 ml
$252.50USD / Each
Showing 1- 12 of 12

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat