Vector Laboratories Normal Sera

Ideal for use in blocking nonspecific binding
  • Supplied undiluted with 0.08% sodium azide as a preservative
  • Applications include immunohistochemistry, immunocytochemistry, and immunofluorescence
Choose from a variety of sera, which when diluted can be used for blocking nonspecific binding or as an antibody diluent. The sera are pooled samples collected from healthy adult goats, horses, chickens, swine, rabbits, cats, and dogs. Each one is heat-treated and centrifuged to remove precipitates and then filtered. The sera are tested with appropriate antibody to check for possible cross-reactivity.

Cat and dog sera can be added to secondary antibody diluent to adsorb potential cross-reactivity with each animal’s tissue.
$140.00 - $176.50USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Description
Availability Pricing
Vector Laboratories S-1000 Normal Goat Serum, concentrated; 20 mL EW-93951-72 Mfr # S-1000-20
COMPARE
20 Normal Goat Serum, concentrated; 20 mL
$165.00USD / Each
Vector Laboratories S-1012-50 Normal Goat Serum, 2.5% solution (v/v); 50 mL EW-93951-73 Mfr # S-1012-50
COMPARE
50 Normal Goat Serum, 2.5% solution (v/v); 50 mL
$144.00USD / Each
Vector Laboratories S-2000 Normal Horse Serum, concentrated; 20 mL EW-93951-74 Mfr # S-2000-20
COMPARE
20 Normal Horse Serum, concentrated; 20 mL
$165.00USD / Each
Vector Laboratories S-2012-50 Normal Horse Serum, 2.5% solution (v/v); 50 mL EW-93951-76 Mfr # S-2012-50
COMPARE
50 Normal Horse Serum, 2.5% solution (v/v); 50 mL
$140.00USD / Each
Vector Laboratories S-4000 Normal Swine Serum, concentrated; 20 mL EW-93951-78 Mfr # S-4000-20
COMPARE
20 Normal Swine Serum, concentrated; 20 mL
$176.50USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat