Vector Laboratories Peroxidase Substrate Kits

Successfully label multiple antigens in same tissue section
  • Dropper bottles simplify handling of chromogens
  • No wait times for mixing and dissolving powders or tablets
These enzyme substrate kits for use with peroxidase detection systems offer a wide variety of applications. Numerous color choices are available for staining antigens in pigmented tissues and melanomas, as well as for localizing multiple antigens in the same tissue section. Viewing options for the substrates include brightfield, fluorescence, darkfield, electron microscopy, and spectral imaging.

The substrates also come in a varying degree of sensitivities. The ImmPACT peroxidase substrates are about 2-4 times more sensitive than the original peroxidase substrate kits.
$224.00 - $642.00USD / Each

12 variations of this product are available.

See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Color Volume (mL) Description
Availability Pricing
Vector Laboratories SK-4100 DAB Substrate Kit, Brown,  300 ml EW-93951-85 Mfr # SK-4100
COMPARE
Brown, gray-black 300 DAB Substrate Kit, Brown, 300 ml
$265.50USD / Each
Vector Laboratories ImmPACT DAB EqV Substrate, 400 ml EW-93951-86 Mfr # SK-4103-400
COMPARE
Brown 400 DAB EqV Substrate, 400 ml
$642.00USD / Each
Vector Laboratories ImmPACT DAB Substrate Kit, Brown,  120 ml EW-93951-87 Mfr # SK-4105
COMPARE
Brown 120 DAB Substrate Kit, Brown, 120 ml
$224.00USD / Each
Vector Laboratories SK-4200 AEC Substrate Kit, Red, 300 ml EW-93951-88 Mfr # SK-4200
COMPARE
Red 300 AEC Substrate Kit, Red, 300 ml
$227.50USD / Each
Vector Laboratories ImmPACT AMEC Red, 120 ml EW-93951-91 Mfr # SK-4285
COMPARE
Red 120 AMEC Red, 120 ml
$226.50USD / Each
Vector Laboratories SK-4400 TMB Substrate Kit, Blue, 300 ml EW-93951-93 Mfr # SK-4400
COMPARE
Blue 300 TMB Substrate Kit, Blue, 300 ml
$230.00USD / Each
Vector Laboratories SK-4600 VIP Substrate Kit, Purple, 300 ml EW-93951-96 Mfr # SK-4600
COMPARE
Purple 300 VIP Substrate Kit, Purple, 300 ml
$226.50USD / Each
Vector Laboratories ImmPACT VIP Substrate, Purple, 120 ml EW-93951-97 Mfr # SK-4605
COMPARE
Purple 120 VIP Substrate, Purple, 120 ml
$226.50USD / Each
Vector Laboratories SK-4700 SG Substrate Kit, Blue-Gray, 300 ml EW-93951-98 Mfr # SK-4700
COMPARE
Blue-gray 300 SG Substrate Kit, Blue-gray, 300 ml
$226.50USD / Each
Vector Laboratories ImmPACT SG Substrate, Blue-gray, 120 ml EW-93951-99 Mfr # SK-4705
COMPARE
Blue-gray 120 SG Substrate, Blue-gray, 120 ml
$230.00USD / Each
Vector Laboratories Vector NovaRed Substrate Kit, Red, 300 ml EW-93952-00 Mfr # SK-4800
COMPARE
Red 300 Substrate Kit, Red, 300 ml
$237.50USD / Each
Vector Laboratories ImmPACT NovaRed Substrate, Red, 120 ml EW-93952-01 Mfr # SK-4805
COMPARE
Red 120 Substrate, Red, 120 ml
$245.50USD / Each
Showing 1- 12 of 12

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat