This product is now discontinued, View Related Products
Vee Gee Brix% (Total Solids) Hand Refractometers
  • Food and beverage testing
  • Largest display available
  • Durable, all-metal design
Brix refractometers show the concentration of the soluble solids content (total solids) of a water-based solution. The soluble solids content is the total of all the solids dissolved in the water such as sugar, salts, proteins, and acids. The measurement reading value is the sum total.
These refractometers are an excellent choice for testing fruit juices, beverages, jams, jellies and syrups. Also very useful for wineries, breweries, fruit harvesting, and food canning/packaging. An ideal choice for testing in conditions with large ambient temperature fluctuations.
Call For Price
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Max Brix Range (%) Resolution
Availability Pricing
Vee Gee 43015 Handheld Brix Refractometer; 0 to 50% EW-02894-99 Mfr # 43015
COMPARE
50 0.5% (Brix)
Call For Price
Vee Gee 43012 Brix Hand Refractometer, 0 to 20% Brix with ATC EW-26048-64 Mfr # 43012
COMPARE
20 0.1% (Brix)
Call For Price
Vee Gee 43003 Handheld Brix Refractometer; 28 to 62% EW-26048-65 Mfr # 43003
COMPARE
62 0.2% (Brix)
Call For Price
Vee Gee 43005 Handheld Brix Refractometer; 58 to 92% EW-26048-66 Mfr # 43005
COMPARE
92 0.2% (Brix)
Call For Price
Vee Gee 43007 Handheld Brix Refractometer; 45 to 82% EW-26048-67 Mfr # 43007
COMPARE
82 0.2% (Brix)
Call For Price
Vee Gee 43001 Handheld Brix Refractometer; 0 to 32% EW-26048-68 Mfr # 43001
COMPARE
32 0.2% (Brix)
Call For Price
Vee Gee 43002 Brix Hand Refractometer, 0 to 32% Brix with ATC EW-26048-69 Mfr # 43002
COMPARE
32 0.2% (Brix)
Call For Price
Vee Gee 43009 Handheld Brix Refractometer; 0 to 10% EW-26048-70 Mfr # 43009
COMPARE
10 0.1% (Brix)
Call For Price
Vee Gee 43017 Handheld Brix Refractometer; 0 to 90% EW-26048-71 Mfr # 43017
COMPARE
90 0.2% (Brix)
Call For Price
Showing 1- 9 of 9