Velp ECO 8 and ECO 25 Thermal Reactors

COD and TNT reactor for EPA-approved digestion methods
  • Perform COD analysis in only 30 minutes at 160°C
  • Ideal for digestion of 16-mm round-bottom test tubes
Velp thermal reactors are PID controlled to offer optimal temperature stability and homogeneity to your reactions. Keypad lets you select from five working temperatures (70, 100, 120, 150, and 160°C) with four time settings (30, 60, 120 minutes, or continuous operation). Units sound an alarm at the end of cycle, and automatically shut down.
$1,011.00 - $1,277.00USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Velp ECO 8 ECO8 Thermoreactor, holds 8 X 16 mm tubes, 115-230 VAC EW-99561-21 Mfr # F101A0127
COMPARE
ECO8 Thermoreactor, holds 8 X 16 mm tubes, 115-230 VAC
$1,011.00USD / Each
Velp ECO 25 ECO25 Thermoreactor, holds 25 X 16 mm tubes, 115-230 VAC EW-99561-31 Mfr # F101A0125
COMPARE
ECO25 Thermoreactor, holds 25 X 16 mm tubes, 115-230 VAC
$1,277.00USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat