Versilon™ 2001 Transfer Tubing

Ideal for chemical transfer applications
  • Global market leader in range of formulations for most extensive chemical compatibility
Formulation: Versilon™ 2001
Applications: Soap and detergent dispensing, water purification lines, food contact applications and chemical transfer.
Characteristics: Plasticizer-free, translucent, excellent chemical compatibility to weak and medium acids and bases, salts and alcohols.
Certification:FDA compliant for food contact
Temperature range:-108F to 135F (-78 to 57C)
Sterilize: With ethylene oxide, gamma irradiation or steam at 30 minutes at 15 psi (1.0 bar).
$122.00 - $1,383.00USD / Pkg of 1

10 variations of this product are available.

See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
Tygon Transfer Tubing, Versilon™ 2001, 1/16" ID x 3/16" OD; 50 Ft
Buy More Save More
EW-96430-00 Mfr # AE300003
COMPARE
45 1/16 3/16
$122.00USD / Pkg of 1
Tygon Transfer Tubing, Versilon™ 2001, 1/8" ID x 1/4" OD; 50 Ft
Buy More Save More
EW-96430-02 Mfr # AE300007
COMPARE
30 1/8 1/4
$182.00USD / Pkg of 1
Tygon Transfer Tubing, Versilon™ 2001, 3/16 x 5/16, 50'
Buy More Save More
EW-96430-04 Mfr # AE300012
COMPARE
22 3/16 5/16
$182.00USD / Pkg of 1
Tygon Transfer Tubing, Versilon™ 2001, 1/4" ID x 3/8" OD; 50 Ft
Buy More Save More
EW-96430-06 Mfr # AE300017
COMPARE
17 1/4 3/8
$231.00USD / Pkg of 1
Tygon Transfer Tubing, Versilon™ 2001, 5/16 x 7/16, 50'
Buy More Save More
EW-96430-08 Mfr # AE300022
COMPARE
14 5/16 7/16
$257.00USD / Pkg of 1
Tygon Transfer Tubing, Versilon™ 2001, 3/8" ID x 1/2" OD; 50 Ft
Buy More Save More
EW-96430-10 Mfr # AE300027
COMPARE
12 3/8 1/2
$286.00USD / Pkg of 1
Tygon Transfer Tubing, Versilon™ 2001, 1/2" ID x 3/4" OD; 50 Ft
Buy More Save More
EW-96430-12 Mfr # AE300038
COMPARE
16 1/2 3/4
$625.00USD / Pkg of 1
Tygon Transfer Tubing, Versilon™ 2001, 5/8" ID x 7/8" OD; 50 Ft
Buy More Save More
EW-96430-14 Mfr # AE300046
COMPARE
13 5/16 7/8
$733.00USD / Pkg of 1
Tygon Transfer Tubing, Versilon™ 2001, 3/4" ID x 1" OD; 50 Ft
Buy More Save More
EW-96430-16 Mfr # AE300053
COMPARE
11 3/4 1
$795.00USD / Pkg of 1
Tygon Transfer Tubing, Versilon™ 2001, 1" ID x 1-3/8" OD; 50 Ft
Buy More Save More
EW-96430-18 Mfr # AE300064
COMPARE
13 1 1
$1,383.00USD / Pkg of 1
Showing 1- 10 of 10

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat