Versilon™ Silver Transfter Tubing

Silver ion antimicrobial coating inhibits microbe growth
  • Global market leader in range of formulations for most extensive chemical compatibility
Formulation: Bio-based, phthalate-free flexible tubing
Applications: Food and beverage processing, dairy, laboratory, environmental, chemical, and water purification.
Characteristics: Silver ion antimicrobial coating inhibits microbe growth; not temperature sensitive. Long life, chemically resistant, nontoxic. Translucent silver.
Certifications: FDA compliant, and meets NSF-listed Standard 51.
Temperature range: — 31 to 165°F ( — 35 to 74°C)
Sterilize: Ethylene oxide.
$181.00 - $492.00USD / Pkg of 1

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
Tygon Transfer Tubing, Versilon™ Silver, 1/8" ID x 1/4" OD; 50 Ft
Buy More Save More
EW-90020-01 Mfr # AS600007
COMPARE
65 1/8 1/4
$181.00USD / Pkg of 1
Tygon Transfer Tubing, Versilon™ Silver, 3/16" ID x 5/16" OD; 50 Ft
Buy More Save More
EW-90020-02 Mfr # AS600012
COMPARE
50 3/16 5/16
$214.00USD / Pkg of 1
Tygon Transfer Tubing, Versilon™ Silver, 1/4" ID x 3/8" OD; 50 Ft
Buy More Save More
EW-90020-03 Mfr # AS600017
COMPARE
40 1/4 3/8
$227.00USD / Pkg of 1
Tygon Transfer Tubing, Versilon™ Silver, 3/8" ID x 1/2" OD; 50 Ft
Buy More Save More
EW-90020-04 Mfr # AS600027
COMPARE
30 3/8 1/2
$329.00USD / Pkg of 1
Tygon Transfer Tubing, Versilon™ Silver, 1/2" ID x 3/4" OD; 50 Ft
Buy More Save More
EW-90020-05 Mfr # AS600038
COMPARE
38 1/2 3/4
$492.00USD / Pkg of 1
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat