Vortex Attachments
Use these vortex attachments to agitate microcentrifuge tubes. Attachments work with any vortex shaker that has a rubber pad or test tube cup. Sliding disc keeps tube caps securely closed during shaking. Unit is constructed of sturdy plastic. Bel-Art.
$154.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Scienceware 37080-0020 Vortex attachment, 1.5 mL, 20 tubes held EW-04723-61 Mfr # 37080-0020
COMPARE
Vortex attachment, 1.5 mL, 20 tubes held
$154.00USD / Each