Walker Fluxgate Magnetometers
Best instrument for precision measurement of low level magnetic fields
  • Superior stability, linearity, and accuracy
  • Single and triaxial models
  • CE Certified-single axis FGM-2 series
  • Traceable to NIST
These magnetometers are ideal for the following applications:
  • Package inspection
  • Classroom demonstrations
  • Residual field measurements (shielding effectiveness)
  • Solenoid/Helmholtz coil calibration
  • Field Mapping
  • Vehicle magnetic signature measurements
  • Measure Earth’s field vector components
  • Magnetic environment monitoring
  • Measure weak fields in rocks
$3,374.00 - $3,588.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Laboratorio Electrofisco 60-B411793A Single Axis Magnetometer w/Longitudinal Probe EW-20001-63 Mfr # 60-B411793A
COMPARE
Single Axis Magnetometer w/Longitudinal Probe
$3,588.00USD / Each
Laboratorio Electrofisco 60-B411793B Fluxgate Magnetometer EW-20001-64 Mfr # 60-B411793B
COMPARE
Fluxgate Magnetometer
$3,374.00USD / Each