Waring Seven-Speed Blenders
Seve-speed blenders feature push-button, SCR control that provides speeds from 3,500 to 22,000 rpm. Blenders include a container, enamel-finish base, lid, and SS blades. The 120 VAC models are UL-listed and have a 6-ft cord with U.S. standard plug.
$842.00 - $946.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Container Material
Availability Pricing
Waring 7012S 7-Speed Blender, 1L, Stainless Steel 120 VAC EW-04244-75 Mfr # 7012S
COMPARE
Stainless Steel
$946.00USD / Each
Waring 7012G 7-Speed Blender, 1L, Glass Container, 120 VAC EW-04244-85 Mfr # 7012G
COMPARE
Glass
$842.00USD / Each