This product is now discontinued, View Related Products
Waring Two-Liter Blenders 2178
Sealed, precision sleeve bearing system extends the life of the blenders
These midsize blenders process up to 2 liters of your sample at 19,000 or 23,000 rpm in seconds. Units include a stainless steel or polycarbonate container, cover, and heavy-duty motor and blades. The 120 VAC models include 6-ft cord with plug; 240 VAC models come with 6-ft cord only.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Container Material Max RPM Power (VAC)
Availability Pricing
Waring HGBSS Two-Speed Blenders, 20,000 or 24,000 rpm; SS; 2-liters; 120V EW-04254-00 Mfr # HGBSS
COMPARE
Stainless Steel 24000 120
Call For Price
Waring HGB550 Two-Speed Blenders, 16,500 or 20,000 rpm; SS; 2-liters; 240V EW-04254-02 Mfr # HGB550
COMPARE
Stainless Steel 20000 240
Call For Price
Waring HGB500 Two-Speed Blenders, 16,500 or 20,000 rpm; PC; 2-liters; 240V EW-04254-32 Mfr # HGB500
COMPARE
Polycarbonate 20000 240