Warren Rupp Air-Operated Double Diaphragm Pumps

Complies for applications in food processing, pharmaceutical, and cosmetic applications.
  • Handle abrasive and shear-sensitive materials
  • Self-priming up to 24 ft of water
  • Deadhead against closed discharge without damage
These Warren Rupp Air-Operated Double Diaphragm Pumps are available in two compliance models. The USDA certified pump is ideal for clean-in-place sanitary installations. The FDA-compliant pumps are ideal for food processing, pharmaceutical, and cosmetic applications.
$3,059.00 - $5,653.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPM) Outlet Connection Wetted Materials
Availability Pricing
Warren Rupp Air-operated Diaphragm Pump, FDA compliant, 1-1/2" Triclamp, 45gpm EW-74440-50 Mfr # T1FB1SDSWTS600
COMPARE
45 1-1/2" Tri-Clamp® 316 Stainless Steel, Santoprene®
$3,059.00USD / Each
Warren Rupp Air-operated Diaphragm Pump, FDA compliant, 45 Max GPM, 1-1/2" Tri-clamp, PTFE EW-74440-55 Mfr # T1FB1SASWTS600
COMPARE
45 1-1/2" Tri-Clamp® 316 Stainless Steel, PTFE, Nitrile
$3,746.50USD / Each
Warren Rupp Air-operated Diaphragm Pump, FDA compliant, 2" Triclamp, 106gpm EW-74440-60 Mfr # T15B1SDSWTS600
COMPARE
106 2" Tri-Clamp® 316 Stainless Steel, Santoprene®
$5,263.00USD / Each
Warren Rupp Air-operated Diaphragm Pump, FDA compliant, 106 Max GPM, 2" Tri-clamp, PTFE EW-74440-65 Mfr # T15B1SSSWTS600
COMPARE
106 2" Tri-Clamp® 316 Stainless Steel, PTFE, Nitrile
$5,653.00USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat