Warren Rupp Surge Suppressors and Dampeners
Minimize pulsation of positive displacement pumps
  • Less vibration and noise
  • Simple installation
  • Variety of materials of construction to meet the system conditions
These Warren Rupp surge suppressors and dampeners are ideal for use to minimize pulsation while pumping with your positive displacement pump. Choose from a wide range of materials based on the chemical compatibility required.
$775.00 - $1,489.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Wetted Materials
Availability Pricing
Warren Rupp DA051PPTNS00 Surge Dampeners, 0.5" NPTF, PP Body, PTFE Diaphragm EW-75008-00 Mfr # DA051PPTNS00
COMPARE
Polypropylene, PTFE
$836.00USD / Each
Warren Rupp DA051PPTNS00 Surge Dampeners, 0.5" NPTF, PVDF Body, PTFE Diaphragm EW-75008-02 Mfr # DA051PPTNS00
COMPARE
PVDF, PTFE
$775.00USD / Each
Warren Rupp DA071PPTNS00 Surge Dampeners, 0.75" NPTF, PP Body, PTFE Diaphragm EW-75008-10 Mfr # DA071PPTNS00
COMPARE
Polypropylene, PTFE
$869.00USD / Each
Warren Rupp DA071KPTNS00 Surge Dampeners, 0.75" NPTF, PVDF Body, PTFE Diaphragm EW-75008-12 Mfr # DA071KPTNS00
COMPARE
PVDF, PTFE
$1,050.00USD / Each
Warren Rupp DA101PPTAS00 Surge Dampeners, 1" Flange, PP Body, PTFE Diaphragm EW-75008-20 Mfr # DA101PPTAS00
COMPARE
Polypropylene, PTFE
$1,026.00USD / Each
Warren Rupp Surge Suppressors, 1" Flange, Aluminum Body, PTFE-Neoprene Diaphragm EW-75008-30 Mfr # TA1-NG1A
COMPARE
Aluminum, PTFE
$1,356.00USD / Each
Warren Rupp Surge Suppressors, 1.5" Flange, Aluminum Body, PTFE-Neoprene Diaphragm EW-75008-40 Mfr # TA1-1/2-NG1A
COMPARE
Aluminum, PTFE
$1,489.00USD / Each
Showing 1- 7 of 7