This product is now discontinued, View Related Products

Water Pump Pressure Switches

Pump control for water systems
  • Suitable for all types of electrically driven water pumps: jets, submersible, reciprocating
  • Direct acting (NC) or reverse acting (NO) available depending on switch type and model number
  • Simple to install - mount in any orientation
Dwyer Series CXA water pump pressure switches have been proven reliable for controlling automatic water systems-ideal for water well pumps and pumping systems. Easily adjust set point and deadband via screws inside the cover.
Call For Price

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI)
Availability Pricing
Dwyer CXA-S1 Water pump pressure switch, NC, range 15-80 psig (1.0-5.5 bar) EW-68063-42 Mfr # CXA-S1
COMPARE
±5 psig (±0.3 bar) 80
Call For Price
Dwyer CXA-S2 Water pump pressure switch, NC, range 30-100 psig (2.1-6.9 bar) EW-68063-44 Mfr # CXA-S2
COMPARE
±5 psig (±0.3 bar) 100
Call For Price
Dwyer CXA-S3 Water pump pressure switch, NC, range 35-150 psig (2.4-10.3 bar) EW-68063-46 Mfr # CXA-S3
COMPARE
±5 psig (±0.3 bar) 150
Call For Price
Dwyer CXA-R1 Water pump pressure switch, NO, range 15-80 psig (1.0-5.5 bar) EW-68063-48 Mfr # CXA-R1
COMPARE
±5 psig (±0.3 bar) 80
Call For Price
Dwyer CXA-R2 Water pump pressure switch, NO, range 30-100 psig (2.1-6.9 bar) EW-68063-50 Mfr # CXA-R2
COMPARE
±5 psig (±0.3 bar) 100
Call For Price
Dwyer CXA-R3 Water pump pressure switch, NO, range 35-150 psig (2.4-10.3 bar) EW-68063-52 Mfr # CXA-R3
COMPARE
±5 psig (±0.3 bar) 150
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat