Whatman 25-mm GD/X Syringe Filters
A unique, built-in prefilter aids in filtering even the most difficult samples
  • Whatman 25-mm GD/X Syringe Filters are designed to facilitate manual filtration of hard-to-filter samples greater than 10 mL
Whatman 25-mm GD/X Syringe Filters contain a unique prefiltration stack of Whatman Multigrade GMF 150 (10:1 µm) and Grade GF/F (0.7 µm) prefilters. Syringe filters feature a polypropylene housing. Multigrade GMF 150 is a combination of two glass microfiber filters in one. Manufactured from 100% borosilicate glass, it is a combination of a coarse layer on top, meshed with a fine layer below.
The Grade GF/F filter will retain fine particles down to 0.7 µm. These prefilters, combined with the available retentate space within the device, allow you to filter even the most difficult samples with less hand force than ever before possible. Compared to the capacity of an unprotected membrane, the volume of sample filtered can be three to seven times greater. Dual magnets in funnel bottom and support base make filter changing quick and easy.
$148.00 - $680.00USD / Pkg of 150
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Pore Size (µm) Sterile
Availability Pricing
Whatman 6901-2502 GD/X 25 mm Syringe Filters, 0.2 um SFCA, Sterile; 50/Pk EW-29705-00 Mfr # 6901-2502
COMPARE
25 0.2 Yes
$259.00USD / Pkg of 50
Whatman 6901-2504 GD/X 25 mm Syringe Filters, 0.45 um SFCA, Sterile; 50/Pk EW-29705-04 Mfr # 6901-2504
COMPARE
25 0.45 Yes
$259.00USD / Pkg of 50
Whatman 6870-2502 25-mm GD/X Syringe Filters, Nylon; pore size, 0.2 µm non-sterile EW-29705-10 Mfr # 6870-2502
COMPARE
25 0.2 No
$612.00USD / Pkg of 150
Whatman 6870-2504 25-mm GD/X Syringe Filters, Nylon; pore size, 0.45 µm non-sterile EW-29705-14 Mfr # 6870-2504
COMPARE
25 0.45 No
$612.00USD / Pkg of 150
Whatman 6878-2502 GD/X 25 mm Syringe Filters, 0.2 um PP; 150/Pk EW-29705-30 Mfr # 6878-2502
COMPARE
25 0.2 No
$612.00USD / Pkg of 150
Whatman 6878-2504 GD/X 25 mm Syringe Filters, 0.45 um PP; 150/Pk EW-29705-32 Mfr # 6878-2504
COMPARE
25 0.45 No
$608.00USD / Pkg of 150
Whatman 6874-2502 GD/X 25 mm Syringe Filters, 0.2 um PTFE; 150/Pk EW-29705-40 Mfr # 6874-2502
COMPARE
25 0.2 No
$612.00USD / Pkg of 150
Whatman 6874-2504 GD/X 25 mm Syringe Filters, 0.45 um PTFE; 150/Pk EW-29705-44 Mfr # 6874-2504
COMPARE
25 0.45 No
$608.00USD / Pkg of 150
Whatman 6900-2502 GD/X 25 mm Syringe Filters, 0.2 um PVDF, Sterile; 50/Pk EW-29705-50 Mfr # 6900-2502
COMPARE
25 0.2 Yes
$231.00USD / Pkg of 50
Whatman 6900-2504 GD/X 25 mm Syringe Filters, 0.45 um PVDF, Sterile; 50/Pk EW-29705-54 Mfr # 6900-2504
COMPARE
25 0.45 Yes
$218.00USD / Pkg of 50
COMPARE
$680.00USD / Pkg
COMPARE
$148.00USD / Pkg
Showing 1- 12 of 12