CYTIVA Whatman™ HEPA-VENT and HEPA-CAP Filters
Preventing bacterical, algal, or fungal contamination in fermentors or incubators
  • Retains 99.97% of airborne particles >=0.3 µm
  • High flow rates with low pressure drop across the filter media
  • Allows bi-directional flow
These in-line filter devices are particularly suitable for the sterile venting of fermentors or reactin vessels to prevent airborne particles from entering or exiting the vessel. Applications include tissue culture, in-line filtration, and removal of particulates form gas/air stream. These lightweight filters have a polypropylene housing and are available in disc and capsule formats. The devices contain glass microfiber filtration media, which is mildly hydrophobic for resistance to bacterial growth and has been laminated on both sides to prevent fiber release. Repeatedly autoclavable for ensured sterility.
CYTIVA Whatman™ Warranties to served market customers are in lieu of all other warranties, expressed or implied, and CYTIVA Whatman™ disclaims all other warranties, including without limitation, the implied warranties of non-infringement, merchantability, fitness for a particular purpose, quiet enjoyment, system integration and data accuracy. CYTIVA Whatman™ limited warranties set forth in the CYTIVA Whatman™ warranty statement shall be enforced by and only by the served market customers.
$100.25 - $159.75USD / Pkg of 1
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Effective Filtration Area (cm²)
Availability Pricing
Whatman 6723-5000 HEPA-VENT Filters; 10pk 50mm Dia
Always in Stock
EW-29700-90 Mfr # 6723-5000
COMPARE
16
$159.75USD / Pkg of 10
Whatman 6702-3600 HEPA-CAP Filter; 36 capsule, 1pk
Always in Stock
EW-29700-92 Mfr # 6702-3600
COMPARE
625
$100.25USD / Pkg of 1
Whatman 6702-7500 HEPA-CAP Filter; 75 capsule, 1pk EW-29700-94 Mfr # 6702-7500
COMPARE
1300
$139.75USD / Pkg of 1