Whatman™ CYTIVA In-Line Solvent Degassers
Simultaneously filter and degas solvents to provide better HPLC reproducibility
  • Eliminate false peaks due to solvent impurities and gas bubbles
Degassers simultaneously filter and degas solvents to provide better HPLC reproducibility. They also eliminate false peaks due to solvent impurities and gas bubbles. Degassers feature built-in 0.2-µm filter. Aqueous phase degasser has a nylon filter and organic phase degasser has a polypropylene filter. Other wetted parts are polypropylene. Each box of degassers includes one set of ferrule nuts for 1/8" OD tubing.
CYTIVA Whatman™ Warranties to served market customers are in lieu of all other warranties, expressed or implied, and CYTIVA Whatman™ disclaims all other warranties, including without limitation, the implied warranties of non-infringement, merchantability, fitness for a particular purpose, quiet enjoyment, system integration and data accuracy. CYTIVA Whatman™ limited warranties set forth in the CYTIVA Whatman™ warranty statement shall be enforced by and only by the served market customers.
$270.50 - $299.75USD / Box of 10
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Whatman In-Line Solvent Degasser, Aqueous Phase; 10/box EW-29702-00 Mfr # 6726-5002
COMPARE
In-Line Solvent Degasser, Aqueous Phase; 10/box
$270.50USD / Box of 10
Whatman In-Line Solvent Degasser, Organic Phase; 10/box EW-29702-10 Mfr # 6725-5002
COMPARE
In-Line Solvent Degasser, Organic Phase; 10/box
$299.75USD / Box of 10