Whatman Quartz Filters

Meet requirements for EPA PM-10 micron and PM-2.5 micron testing
  • Ideal for air particle monitoring and environmental testing
Quartz Filters are used for air sampling of acidic gases, stacks, flues, and aerosols. Effective at high temperatures up to 964°F (500°C). Meet requirements for EPA PM-10 µm and PM-2.5 µm testing to sample particles down to 2.5 µm. High purity quartz (SiO2) microfiber filters have low levels of alkaline earth metals, eliminating artifact products of sulfates and nitrates (from SO2 and NO2).
$171.00 - $403.00USD / Pkg of 100

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Whatman 1851-037 Quartz Filters, QM-A, 3.7 cm Dia EW-29706-10 Mfr # 1851-037
COMPARE
Quartz Filters, QM-A, 3.7 cm Dia
$171.00USD / Pkg of 100
Whatman 1851-047 Quartz Filters, QM-A, 4.7 cm Dia EW-29706-12 Mfr # 1851-047
COMPARE
Quartz Filters, QM-A, 4.7 cm Dia
$187.00USD / Pkg of 100
Whatman 1851-090 Quartz Filters, QM-A, 9.0 cm Dia EW-29706-16 Mfr # 1851-090
COMPARE
Quartz Filters, QM-A, 9.0 cm Dia
$390.00USD / Pkg of 100
Whatman 1851-865 Quartz Filters, QM-A, 8"W x 10"H EW-29706-22 Mfr # 1851-865
COMPARE
Quartz Filters, QM-A, 8"W x 10"H
$403.00USD / Pkg of 25
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat