This product is now discontinued, View Related Products
Wheaton Economy Sample Bottles
Large writing area allows for easy identification
These economy bottles are manufactured with USP type I borosilicate glass to maintain purity of contents. Special lip and sloping shoulders make pouring and cleaning easy.
Sample bottles are graduated to approximate volume; label space allows clear identification. Bottles include chemical-resistant, PTFE-lined screw caps (size 33 mm).Bottles and caps are autoclavable.
$349.00 - $534.00USD / Case of 48
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
DWK Life Sciences (Wheaton) 219817 Economy Sample Bottle; Graduated, 250 mL, 48/cs EW-08913-25 Mfr # 219817
COMPARE
250 25 Glass
$534.00USD / Case of 48
DWK Life Sciences (Wheaton) 219819 Economy Sample Bottle; Graduated, 500 mL, 24/cs EW-08913-45 Mfr # 219819
COMPARE
500 50 Glass
Call For Price
DWK Life Sciences (Wheaton) 219820 Economy Sample Bottle; Graduated, 1 L, 24/cs EW-08913-55 Mfr # 219820
COMPARE
1000 50 Glass
$349.00USD / Case of 24