Wheaton Socorex Calibrex organo 525 Bottletop Dispensers
Three volume ranges to cover your dispensing needs!
  • Built to be autoclaved fully assembled at 250°F (121°C) for 20 minutes
  • Integrated calibration system for easy in-house adjustments
  • Chemically inert and transparent fluid path
The organo 525 model features a ground glass plunger, making it ideal for organics, and noncrystallizing acid and base solutions. A built-in calibration system, hidden under the cap, allows for simple adjustment to accurately accommodate solutions due to varying viscosity or temperature. Two volume adjustment knobs are included—use the premounted slider when changing dispensing volumes frequently or the screw-type version for a more permanent setting without the risk of accidental changes.
$1,077.00 - $1,663.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Division (mL) Max Volume (mL) Min Volume (mL)
Availability Pricing
DWK Life Sciences (Wheaton) Organo 525 Calibrex 525 organo Bottletop Dispenser, 2.5 to 25 mL EW-07914-46 Mfr # W844090
COMPARE
0.5 25 2.5
$1,077.00USD / Each
DWK Life Sciences (Wheaton) Organo 525 Calibrex 525 organo Bottletop Dispenser, 5 to 50 mL EW-07914-47 Mfr # W844092
COMPARE
1 50 5
$1,104.00USD / Each
DWK Life Sciences (Wheaton) Organo 525 Calibrex 525 organo Bottletop Dispenser, 10 to 100 mL EW-07914-48 Mfr # W844094
COMPARE
1 100 10
$1,663.00USD / Each