Wika C10 OEM Pressure Transmitters
Rugged design withstands high shock, vibration, and moisture
These WIKA pressure transmitters and transducers are precision engineered to meet the price and performance requirements of Original Equipment Manufacturers. Each unit undergoes extensive quality control testing and calibration to achieve an accuracy < 0.50% full scale. The printed circuit boards use state-of-the-art surface mount technology and are potted in silicone gel for protection against mechanical shock, vibration, and moisture. Each unit is temperature compensated to assure accuracy and long- term stability when exposed to severe ambient temperature variations.
They provide a rugged design featuring excellent vibration resistance and long service life. OEM applications include hydraulics, pneumatics, press control, compressor control, pump protection and numerous other process
$440.87USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Description
Availability Pricing
Wika C-10 Pressure Transmitter, 0 to 50 psi EW-68446-92 Mfr # 8363468
COMPARE
±0.5% BFSL 50 Pressure Transmitter, 0 to 50 psi
$440.87USD / Each
Wika C-10 Pressure Transmitter, 0 to 300 psi EW-68446-95 Mfr # 8363506
COMPARE
±0.5% BFSL 300 Pressure Transmitter, 0 to 300 psi
$440.87USD / Each