Wika Industrial Stainless Steel Gauges - Type 232.54
These gauges feature 316 stainless steel wetted parts and 304 stainless steel case and bayonet ring, an adjustable pointer, a laminated safety glass window, and can be liquid-filled in the field.
WIKA stainless steel gauges are ideal for process, chemical, and petrochemical applications, oil exploration and production, power generation, pollution control equipment, and any other application requiring fine quality, long lasting pressure measurement instrumentation.
  • Center Back Mount Connection Available
  • 1/4" NPT Connection Size
  • 4" Dial Size Available
$68.25 - $506.00USD / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Dial Size (in) Max Pressure (PSI)
Availability Pricing
Wika 232.54 2.5 Industrial Stainless Steel Gauge, 0 to 30 psi EW-68432-57 Mfr # 9744908
COMPARE
±2-1-2% full-scale 2.5 30
$72.75USD / Each
Wika 232.54 2.5 Industrial Stainless Steel Gauge, 0 to 160 psi EW-68432-58 Mfr # 9744932
COMPARE
±2-1-2% full-scale 2.5 160
$72.75USD / Each
Wika 232.54 2.5 Industrial Stainless Steel Gauge, 0 to 200 psi EW-68432-59 Mfr # 9744940
COMPARE
±2-1-2% full-scale 2.5 200
$72.75USD / Each
Wika 232.54 2.5 Industrial Stainless Steel Gauge, 0 to 300 psi EW-68432-60 Mfr # 9744959
COMPARE
±2-1-2% full-scale 2.5 300
$72.75USD / Each
Wika 232.54 2.5 Industrial Stainless Steel Gauge, 0 to 600 psi EW-68432-62 Mfr # 9744975
COMPARE
±2-1-2% full-scale 2.5 600
$72.75USD / Each
Wika 232.54 2.5 Industrial Stainless Steel Gauge, 0 to 1000 psi EW-68432-63 Mfr # 9744991
COMPARE
±2-1-2% full-scale 2.5 1000
$72.75USD / Each
Wika 232.54 2.5 Industrial Stainless Steel Gauge, 0 to 1500 psi EW-68432-64 Mfr # 9745009
COMPARE
±2-1-2% full-scale 2.5 1500
$78.75USD / Each
Wika 232.54 2.5 Industrial Stainless Steel Gauge, 0 to 10,000 EW-68432-66 Mfr # 9745041
COMPARE
±2-1-2% full-scale 2.5 10000
$78.75USD / Each
Wika 232.54 2.5 Industrial Stainless Steel Gauge, 30" to 0" Hg EW-68432-71 Mfr # 9744827
COMPARE
±2-1-2% full-scale 2.5 14.375
$68.25USD / Each
Wika 232.54 2.5 Industrial Stainless Steel Gauge, 0 to 15 psi EW-68432-72 Mfr # 9744894
COMPARE
±2-1-2% full-scale 2.5 15
$72.75USD / Each
Wika 232.54 2.5 Industrial Stainless Steel Gauge, 0 to 60 psi EW-68432-73 Mfr # 9744916
COMPARE
±2-1-2% full-scale 2.5 60
$72.75USD / Each
Wika 232.54 2.5 Industrial Stainless Steel Gauge, 0 to 100 psi EW-68432-74 Mfr # 9744924
COMPARE
±2-1-2% full-scale 2.5 100
$72.75USD / Each
Wika 232.54 2.5 Industrial Stainless Steel Gauge, 0 to 2000 psi EW-68432-75 Mfr # 9745017
COMPARE
±2-1-2% full-scale 2.5 2000
$78.75USD / Each
Wika 232.54 2.5 Industrial Stainless Steel Gauge, 0 to 3000 psi EW-68432-76 Mfr # 9745025
COMPARE
±2-1-2% full-scale 2.5 3000
$78.75USD / Each
Wika GAUGE 6IN 5000PSI 1/2 B FF EW-68433-56 Mfr # 4214421-0001
COMPARE
$506.00USD / Each
Showing 1- 15 of 15